Learn More
Medchemexpress LLC Xanthan gum | 11138-66-2 | 500 G
SDP
Supplier:  Medchemexpress LLC HYA0103500G
Encompass_Preferred
Xanthan gum increases hydrogel viscosity and stability by interacting with gelatin through hydrogen bonds, which promotes cell growth and creates a favorable microenvironment for cell differentiation. It also induces pro-inflammatory responses by elevating TNF-α, IL-6, and IL-10 levels, making it useful for inflammation and immunology research.
  • Increases hydrogel viscosity and stability
  • Promotes cell growth and differentiation
  • Induces pro-inflammatory responses by elevating TNF-α, IL-6, and IL-10 levels
  • Supports cell viability and moisture retention in hydrogels
  • Modulates gut microbiota and reduces C. difficile colonization
Catalog No. 50-004-32188