Learn More
Medchemexpress LLC Xanthine-13C,15N2 | 1262670-81-4 | MFCD34676312 | 98.0% | 155.09 | C4[13C]H4N2[15N]2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW017389S10MG
Encompass_Preferred
Xanthine-13C,15N2 is a stable isotope-labeled form of xanthine (one 13C and two 15N labels) supplied as a 10 mg vial for research use. It is used as an internal standard and tracer for quantitative mass spectrometry, metabolite profiling, and metabolic pathway studies. Consult the product datasheet for lot-specific certificates and handling instructions.
  • Stable isotope labeled for mass spectrometry internal standard use.
  • Contains one 13C and two 15N labels for unambiguous isotopic tracking.
  • Supplied as a 10 mg vial suitable for analytical applications.
  • High purity consistent with analytical workflows.
  • Intended for research use; not for human or clinical use.
Catalog No. 50-002-99949