Learn More
Medchemexpress LLC Xanthocillin X permethyl ether | 4464-33-9 | 98.91% | 316.35 | 50 MG

Supplier: Medchemexpress LLC HY11191150MG
Xanthocillin X permethyl ether is a natural compound isolated from fungal extracts, known for its Aβ-42 lowering activity. This compound is suitable for various laboratory applications.
- Isolated from fungal extracts
- Exhibits Aβ-42 lowering activity
- High purity of 98.91%
- Solid appearance, light yellow to yellow color
- Soluble in DMSO (5 mg/mL)
- Structure classified as an antibiotic, antifungal, and for disease research
- Initial source: microorganisms, Basipetospora sp.
- Storage conditions: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.