Learn More
ATCC Xanthomonas campestris (Pammel) Dowson, strain BB-1 - freeze-dried culture (ATCC 31922)
Supplier:  ATCC 31922
Encompass
Freeze-dried culture of Xanthomonas campestris (Pammel) Dowson, strain BB-1, intended for research and laboratory characterization. Supplied as a freeze-dried vial for reconstitution and cultivation under standard microbiological procedures. Handle according to institutional biosafety guidelines.
  • Freeze-dried format for long-term stability.
  • Strain designation BB-1 for characterization and comparative studies.
  • Suitable for laboratory research and culture propagation.
  • Recommended growth temperature 28 °C.
  • Biosafety level 1; follow institutional biosafety procedures.
  • Supplied as a single vial for reconstitution.
Catalog No. NC3866623