Learn More
Selleck Chemical LLC Xanthosine S9369-10MG
SDP
Supplier:  Selleck Chemical LLC S936910MG
Encompass_Preferred
Xanthosine (Xanthine riboside) is an intermediate in purine metabolism formed from IMP and forming GMP
Catalog No. 50-000-24557