Learn More
Medchemexpress LLC Xanthotoxol | 2009-24-7 | 99.5% | 202.16 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY30152100MG
Encompass_Preferred
Xanthotoxol (8-Hydroxypsoralen) is a fragrant bean substance and a CYP450 inhibitor. It exhibits anti-inflammatory, and 5-HT antagonistic and protective effects. Xanthotoxol inhibits CYP3A4 and CYP1A2 with IC50s of 7.43 μM and 27.82 μM, respectively. It can pass through MAPK and NF-κB pathways, inhibiting inflammation.
  • Fragrant bean substance
  • CYP450 inhibitor
  • Anti-inflammatory effects
  • 5-HT antagonistic and protective effects
  • Inhibits CYP3A4 and CYP1A2
  • Acts through MAPK and NF-κB pathways to inhibit inflammation
Catalog No. 50-004-05235