Learn More
Apexbio Technology LLC Xanthurenic acid 59-00-7 50mg
SDP
Supplier:  Apexbio Technology LLC B759350
Encompass_Preferred
Xanthurenic acid (CAS 59-00-7) is a metabolite derived from the tryptophan catabolic pathway It has been shown to modulate central nervous system activity through selective agonism of group II metabotropic glutamate receptors (mGluR II) particularly in the hypothalamic region This interaction influences neuronal signal transduction within the hypothalamus Xanthurenic acid is primarily utilized in research aimed at elucidating the roles of glutamatergic signaling pathways in sensory transmission and neural regulation as well as broader neurobiological studies involving neurotransmission and central nervous system physiology
Catalog No. 50-000-91746