Learn More
Medchemexpress LLC Xanthurenic acid | 59-00-7 | MFCD00006754 | 99.4% | 205.17 g/mol | C10H7NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW014666R5MG
Encompass_Preferred
Xanthurenic acid (CAS 59-00-7) is a tryptophan metabolite provided as a high-purity analytical reference material for research and method development. It is used in biochemical assays, chromatography, and mass spectrometry to support identification, quantification, and calibration.
  • High purity suitable for analytical and reference applications.
  • Solid form for straightforward storage and handling.
  • Compatible with HPLC and mass spectrometry workflows.
  • Useful for method development, calibration, and identification studies.
  • Supplied in small vials appropriate for low-quantity analytical work.
Catalog No. 50-003-02924