Learn More
Medchemexpress LLC Xanthurenic acid | 59-00-7 | MFCD00006754 | 99.5% | 205.17 g·mol⁻1 | C10H7NO4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0146665G
Encompass_Preferred
Xanthurenic acid is an endogenous metabolite reported as a putative Group II metabotropic glutamate receptor (mGluR II) agonist. It is supplied for research use and as an analytical standard for chromatographic and mass spectrometric methods, and is typically offered at high purity for quantitative and qualitative studies.
  • Endogenous metabolite and putative Group II metabotropic glutamate receptor agonist.
  • Supplied as a research-grade compound and analytical standard for HPLC, GC, and MS.
  • High purity suitable for quantitative and qualitative analysis.
  • Available in multiple pack sizes from milligram to gram scale.
  • Off-white to light yellow solid, convenient for handling and storage.
  • Molecular formula C10H7NO4; molecular weight 205.17 g·mol⁻1.
Catalog No. 50-003-02426