Learn More
WELCH MATERIALS INC Ultisil XB-C18, 5µm, 21.2×250mm
Supplier:  WELCH MATERIALS INC H0261021106
Encompass_Preferred
Prep Column, Ultisil XB-C18, 5µm, 21.2×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052573