Learn More
WELCH MATERIALS INC Ultisil XB-C4, 5µm, 300Å, 4.6×250mm
Supplier:  WELCH MATERIALS INC H0021633043
Encompass_Preferred
Analytical Column, Ultisil XB-C4, 5μm, 300Å, 4.6×250mm, pH range: 1.5-10.0, Pore Size: 300Å, USP listing: L26, USP listing: L26, Carbon Loading: 3%, Surface Area: 90m2/g, endcapped.
Catalog No. NC2052226