Learn More
WELCH MATERIALS INC Ultisil XB-C8, 20-40µm, 100Å, 1kg
Supplier:  WELCH MATERIALS INC H0272014600
Encompass_Preferred
Bulk Media, Ultisil XB-C8, 20-40μm, pH range: 1.5-10.0, Pore Size: 100Å, USP listing: L7, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped. Packing: 1kg/bottle.
Catalog No. NC2052752