Learn More
WELCH MATERIALS INC Ultisil XB-NH2, 40-70µm, 100Å, 1kg
Supplier:  WELCH MATERIALS INC H0273011600
Encompass_Preferred
Bulk Media, Ultisil XB-NH2, 40-70μm, pH range: 2.0-8.0, Pore Size: 100Å, USP listing: L8, Carbon Loading: 4%, Surface Area: 320m2/g, not-endcapped. Packing: 1kg/bottle.
Catalog No. NC2052757