Learn More
CLICKBIO INC XBLOK 650 E-Beamed Irradiated (10/CS)
Supplier:  CLICKBIO INC XBLOK6501E
Encompass
The XBLOK650 is a polypropylene ANSI-SLAS footprint plate with 6 w x 50 mL. The well shape is based on common 50 mL conical tubes. Ideal for large volume sample prep, tissue homogenization, or suspension cell culture workflows. It is e-beam irradiated for aseptic workflows.
Catalog No. NC2383506