Learn More
CLICKBIO INC XBLOK 2412 Plate (20/CS)
Supplier:  CLICKBIO INC CBXBLOK24121
Encompass
The XBLOK2412 is a polypropylene ANSI-SLAS footprint plate with 24 x 12 mL wells. It is ideal for parallel fermentation and sample preparation workflows.
Catalog No. NC2948140