Learn More
Waters Corp XBridge BEH Amide Column, 130Å, 3.5 µm, 3 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186004863
Encompass_Preferred
XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars. XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2-11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa.
Catalog No. 50-791-216