Learn More
Waters Corp Waters XBridge BEH Amide, 130Å, 2.5 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 186006724
Encompass_Preferred
XBridge BEH Amide columns are designed to retain extremely polar compounds like carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, which maintains stability from pH 2 to 11. These columns offer exceptional separation performance, robustness, and high throughput for HPLC assays, and provide a seamless transition path towards UPLC adoption.
  • Chemically stable, trifunctionally bonded amide phase for pH 2 to 11 stability.
  • Enables separation of polar analytes with varying polarity and pKa.
  • Packed in ultra-low dispersion hardware for enhanced performance.
  • 2.5 μm particle technology delivers superior separation and throughput for HPLC and UPLC.
  • Offers excellent chemical stability at high pH and temperatures.
  • BEH particle technology improves hydrolytic stability and particle strength, reducing LC/MS leakage.
Catalog No. 50-466-432