Learn More
Waters Corp Waters XBridge BEH Amide Method Validation Kit, 130Å, 2.5 μm, 3 mm X 150 mm
Supplier:  Waters Corp 186006783
Encompass_Preferred
This method validation kit consists of three columns packed with three different batches of sorbent. It is designed to retain extremely polar compounds, including carbohydrates and sugars. The chemically stable, trifunctionally bonded amide phase is stable from pH 2 to 11, enabling the separation of polar analytes spanning a wide range of polarity, structural moiety, and pKa.
  • Packed in ultra-low dispersion hardware (2.5 μm eXtended Performance [XP] columns).
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-466-475