Learn More
Waters Corp XBridge BEH Amide XP Column, 130Å, 2.5 µm, 4.6 mm X 30 mm, 1/pk
Supplier:  Waters Corp 186006096
Encompass_Preferred
XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars. XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-201-534