Learn More
Waters Corp Waters Column, Amide, 2.5 μm, 2.1 mm, 100 mm
Supplier:  Waters Corp 176002589
Encompass_Preferred
XBridge BEH Amide columns retain extremely polar compounds like carbohydrates and sugars, utilizing a chemically stable, trifunctionally bonded amide phase with pH stability from 2 to 11. These 2.5 μm “eXtended Performance” columns, packed in ultra-low dispersion hardware, deliver exceptional separation, robustness, and throughput for HPLC, and offer a seamless transition to UPLC. Their particle technology enhances hydrolytic stability and strength, preventing LC/MS leakage.
  • Highly customizable for designing robust procedures using various mobile phases with a wide pH range.
  • Allows for generating samples with outstanding chemical stability at high pH and temperatures, even with increased mass loading of basic chemicals.
Catalog No. 50-179-248