Learn More
Waters Corp XBridge BEH Amide Column, 130Å, 2.5 μm, 4.6 mm X 100 mm
Supplier:  Waters Corp 186006099
Encompass_Preferred
XBridge BEH Amide columns are designed for retaining extremely polar compounds like carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11, facilitating separation of diverse polar analytes. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, offering exceptional performance, robustness, and throughput for HPLC assays and enabling smooth UPLC adoption.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase stable from pH 2 to 11.
  • Packed in ultra-low dispersion hardware (2.5 μm eXtended Performance [XP] columns).
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • BEH particle technology improves hydrolytic stability and particle strength, preventing LC/MS leakage.
Catalog No. 50-201-537