Learn More
Waters Corp XBridge Peptide BEH C18 Column, 300Å, 2.5 µm, 2.1 mm × 150 mm
Supplier: Waters Corp 186009079
This (U)HPLC column kit features BEH C18 chemistry, 300 Å pore size, and 2.5 µm particle size for analytical chromatography. Made with stainless steel hardware, it offers robust performance for various laboratory applications.
- Length: 150 mm
- Internal diameter: 2.1 mm
- Particle size: 2.5 µm
- Chemistry: C18 (RP-18, ODS, Octadecyl)
- Pore size: 300 Å
- USP class: L1
- Carbon load: 12.0 %
- Endcapping: endcapped
- Column type: Analytical column
- Hardware material: Stainless steel
- pH range: 1.0 to 12.0
- Temperature range: 60 °C to 80 °C
- Mode of separation: Reversed phase chromatography (RP)
- Technology: U(H)PLC
- Matrix: Hybrid
- Particle morphology: spherical, fully porous
- Pressure stability: 415 bar
- Packaging unit: 3 pcs
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.