Learn More
Waters Corp XBridge BEH C18 OBD Prep Column, 10 μm, 10 mm × 250 mm
Supplier:  Waters Corp 186008211
Encompass_Preferred
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Broad pH range (1 to 12) for method development.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Superior low pH stability extends column life.
  • Ultra-low column bleed improves detection sensitivity.
  • Optimum Bed Density (OBD) design resists mechanical failure and pressure shocks.
  • Increased productivity through faster lab-scale separations and seamless scaling.
  • Longer column lifetimes reduce replacement frequency.
Catalog No. 50-239-2350