Learn More
Waters Corp Waters Column, C18, 2.5 μm, 130 Å, 3 mm x 100 mm
Supplier:  Waters Corp 186008983
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column is optimized and QC tested for peptide separations. Featuring a small-pore (130Å) trifunctionally bonded BEH particle, it offers a wide usable pH range, superior low pH stability, and ultra-low column bleed, making it suitable for proteomic, peptide mapping, and synthetic peptide applications.
  • Optimized and QC tested for peptide separations, ensuring reliability.
  • Features small-pore (130Å) trifunctionally bonded BEH particles for broad pH compatibility.
  • Provides superior low pH stability and ultra-low column bleed.
  • Delivers excellent peak shape, high efficiency, and outstanding stability.
  • Manufactured in a cGMP, ISO 9002 accredited plant using ultra-pure reagents.
  • Includes a Performance Test Chromatogram and Certificate of Acceptance.
  • Carries a USP Classification of L1.
Catalog No. 50-152-8606