Learn More
Waters Corp Waters Column, BEH C18, 2.5 μm, 3 × 75 mm
Supplier:  Waters Corp 176002552
Encompass_Preferred
The XBridge BEH C18 sorbent is ideal for HPLC method development, offering a broad mobile phase pH range of 1 to 12. Its C18 ligand provides superior low pH stability and ultra-low column bleed. These 2.5 μm XP columns ensure exceptional separation performance, robustness, and throughput for HPLC assays, and enable a seamless transition to UPLC.
  • Operates across a broad 1 to 12 pH range.
  • Provides enhanced stability in acidic conditions.
  • Minimizes stationary phase shedding.
  • Delivers high-quality separations.
  • Offers reliable performance and efficient sample processing for HPLC assays.
  • Allows for easy adoption into UPLC systems.
  • Achieves exceptional peak shape and peak capacity by increasing particle efficiency.
  • Ensures remarkable column longevity.
  • Performs well under high pH settings.
Catalog No. 50-179-211