Learn More
Waters Corp Waters Column, C8, 130Å, 2.5 μm, 3 mm X 150 mm
Supplier:  Waters Corp 186006713
Encompass_Preferred
This column utilizes XBridge BEH C8 sorbent, offering enhanced speed, peak shape, and chromatographic performance for method development. Its trifunctionally bonded sorbent provides a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC applications.
  • Consistently accurate results due to robust column chemistry.
  • Improved pH stability, long column lifetime, and enhanced reliability.
  • Maximizes particle efficiency with unmatched peak shape and peak capacity.
Catalog No. 50-466-421