Learn More
Waters Corp Waters Column, C8, 130Å, 2.5 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186006043
Encompass_Preferred
This BEH C8 sorbent column offers improved peak shape and chromatographic performance with a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed. Designed for exceptional separation performance, robustness, and throughput in HPLC assays, it also facilitates a seamless transition to UPLC.
  • Less retentive for faster method development.
  • Improved peak shape and chromatographic performance.
  • Wide usable pH range (1 to 12) with superior low pH stability.
  • Ultra-low column bleed for reliable results.
  • Exceptional separation performance, robustness, and throughput.
  • Seamless transition path to UPLC adoption.
  • Unrivaled reproducibility, batch to batch and year to year.
  • Longer column lifespan and improved reliability.
  • Maximizes particle efficiency and provides unrivaled peak form and capacity.
Catalog No. 50-179-477