Learn More
Waters Corp Waters Corporation Column, XBridge BEH C8 XP, 130Å, 2.5 μm
Supplier:  Waters Corp 176002561
Encompass_Preferred
XBridge BEH C8 sorbent offers less retention than C18, ideal for faster method development and improved chromatographic performance. The trifunctionally bonded sorbent provides a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns ensure exceptional separation, robustness, and throughput for HPLC assays, aiding UPLC adoption.
  • Less retentive than BEH C18 sorbent, preferred for faster method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent offers the widest usable pH range (1 to 12).
  • Provides superior low pH stability.
  • Features ultra-low column bleed.
  • Ultra-low dispersion hardware ensures exceptional separation, robustness, and throughput for HPLC assays.
  • Facilitates seamless transition to UPLC adoption.
Catalog No. 50-179-220