Learn More
Waters Corp Column, C8, 2.5 μm, 130 Å, 3 mm X 75 mm
Supplier:  Waters Corp 186006046
Encompass_Preferred
The XBridge BEH C8 sorbent has less retention compared to the XBridge BEH C18 sorbent. It is suitable for methods requiring speed, improved peak shape, and enhanced chromatographic performance. The sorbent is trifunctionally bonded, offering a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to BEH C18 sorbent.
  • Offers speed for method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-179-480