Learn More
Waters Corp Waters HPLC Column, XBridge BEH C8, 2.5 μm, 2.1 mm × 50 mm, 130 Å
Supplier:  Waters Corp 186006041
Encompass_Preferred
The XBridge BEH C8 sorbent offers faster separations and improved chromatographic performance compared to C18. It provides a wide usable pH range (1–12), superior low pH stability, and ultra-low column bleed. These 2.5 μm eXtended Performance (XP) columns deliver exceptional separation, robustness, and throughput for HPLC assays, enabling a smooth transition to UPLC.
  • Compatible with all HPLC, UHPLC, and UPLC technology platforms.
  • Achieves exceptional performance and equivalent backpressure to core-shell columns.
  • Offers improved loading capacity and particle size scalability inherent to fully porous particle columns.
  • Based on Ethylene Bridged Hybrid (BEH) particle for enhanced performance and lifetime under harsh conditions.
  • Each batch chromatographically tested to ensure reproducible performance.
Catalog No. 50-179-475