Learn More
Waters Corp Waters Corporation XBridge BEH C8 XP Column, 2.5 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 176002557
Encompass_Preferred
The XBridge BEH C8 XP Column is a high-performance liquid chromatography column featuring BEH C8 chemistry with a 2.5 μm particle size. It offers exceptional separation performance, robustness, and a wide usable pH range of 1 to 12. Designed to maximize productivity in developing LC/MS assays and quality control methods, it facilitates a seamless transition to UPLC adoption.
  • Less retentive sorbent for faster method development.
  • Wide usable pH range of 1 to 12 with superior pH stability.
  • Ultra-low column bleed ensures clean baselines.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Maximized particle efficiency for unmatched peak shape and capacity.
  • Unmatched reproducibility for consistent results.
  • Recommended operating temperature range: 20°C to 80°C.
Catalog No. 50-179-216