Learn More
Waters Corp XBridge BEH C8 XP Column, 130Å, 2.5 µm, 4.6 mm X 75 mm, 1/pk
Supplier:  Waters Corp 186006050
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-201-520