Learn More
Waters Corp Waters XBridge HILIC Column, 130Å, 2.5 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186006079
Encompass_Preferred
XBridge BEH HILIC columns feature an unbonded BEH substrate, engineered for the retention and separation of highly polar compounds with efficient and reproducible HPLC HILIC separations. The BEH particle's chemical stability ensures extended column lifetimes, outperforming silica-based HILIC phases. These columns offer alternate selectivity to standard reversed-phase columns, providing superior peak shape, retention, and longevity.
  • Optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • Exceptional separation performance, robustness, and throughput for HPLC assays with 2.5 μm eXtended Performance [XP] columns.
  • Enables a seamless transition path toward UPLC adoption.
  • pH range of 1 to 9.
  • Withstands operational pressures up to 18000 psi (1240 Bar).
  • Helps retain extremely polar species difficult to keep in regular reversed-phase chromatography.
Catalog No. 50-179-501