Learn More
Waters Corp XBridge BEH HILIC XP Column, 130Å, 2.5 μm, 2.1 mm x 75 mm
Supplier:  Waters Corp 176002580
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • Ethylene Bridged Hybrid (BEH) particle technology ensures maximum column performance and extended column lifetimes.
  • Offers advantages over conventional silica-based particles, with controlled silanol activity for better reproducibility, peak shape, and efficiency.
  • Sets the performance standard for method development.
  • Allows seamless transfer from analytical to preparative separations.
  • General purpose HILIC column.
  • Alternative selectivity to BEH Amide chemistries.
  • Unique selectivity for strong polar basic compounds.
  • Excellent chemical stability from pH 1–9.
Catalog No. 50-179-239