Learn More
Waters Corp XBridge Column, 130Å, 2.5 μm, 3 mm × 30 mm
Supplier:  Waters Corp 176002582
Encompass_Preferred
These XBridge BEH HILIC columns are designed for retaining and separating very polar compounds, offering efficient and reproducible results under HPLC HILIC conditions. They provide mechanical strength and chemical stability, ensuring maximum performance and extended lifetimes.
  • Unbonded BEH substrate for polar compound retention.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • BEH construction for mechanical strength and chemical stability.
  • Ethylene Bridged Hybrid technology enhances column performance and longevity.
  • Controls silanol activity for improved reproducibility and peak shape.
  • BEH particles are highly stable, pH resistant, and mechanically strong.
  • Seamless transfer from analytical to preparative separations.
  • Alternative selectivity for strong polar basic compounds.
  • Excellent chemical stability across pH 1–9.
Catalog No. 50-179-241