Learn More
Waters Corp XBridge BEH HILIC Method Validation Kit, 130Å, 2.5 µm, 2.1 mm X 100 mm, 3/pk
Supplier:  Waters Corp 186006222
Encompass_Preferred
This method validation kit consists of three batches of XBridge BEH HILIC columns packed with three different batches of sorbent
Catalog No. 50-179-593