Learn More
Waters Corp Waters Corporation XBridge BEH Phenyl XP Column, 130Å, 2.5 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186006067
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This Phenyl sorbent is chemically stable, providing ultra-low column bleed, long lifetimes, and excellent peak shape. The 2.5 μm XP columns provide exceptional separation performance and robustness for HPLC assays, facilitating a seamless transition to UPLC adoption.
  • Complementary selectivity to C18 stationary phases, useful for analytes with an aromatic ring.
  • Chemically stable Phenyl sorbent commercially available.
  • Ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • eXtended Performance (XP) columns provide exceptional separation performance and robustness.
  • Wide pH range from 1 to 12 pH for robust method development.
  • BEH particle technology enables remarkable low and high pH stability.
Catalog No. 50-179-493