Learn More
Waters Corp Waters Corporation XBridge BEH Phenyl XP Column, 2.5 μm, 3.0 x 50 mm
Supplier:  Waters Corp 186006069
Encompass_Preferred
The XBridge BEH Phenyl XP 2.5 μm, 3.0 x 50 mm column is designed for forensic toxicology, analyzing a comprehensive panel of pain management drugs. It offers alternative selectivity and enhances opiate retention, reducing ion suppression. It achieves excellent baseline separation of isomers and 35 compounds in under 4 minutes, compatible with any HPLC system.
  • Provides alternative selectivity and phenyl functionality.
  • Enhances the retention of opiate compounds.
  • Potentially reduces ion suppression from urinary matrix components.
  • Achieves excellent baseline separation between isomers.
  • Separates an entire panel of 35 compounds in under 4 minutes.
  • Compatible with any HPLC system.
  • Demonstrates excellent separation of amines, PCP, and BZE.
  • Achieves good peak shape for benzodiazepine chromatography.
Catalog No. 50-179-495