Learn More
Waters Corp XBridge BEH Phenyl XP Column, 130Å, 2.5 µm, 2.1 mm X 75 mm, 1/pk
Supplier:  Waters Corp 186006066
Encompass_Preferred
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-492