Learn More
Waters Corp XBridge BEH Phenyl XP Column, 2.5 μm, 130 Å, 3 mm x 50 mm
Supplier: Waters Corp 176002575
XBridge BEH Phenyl XP columns provide complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. These columns offer superior performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC adoption.
- Complementary selectivity to C18 phases for polyaromatic compounds
- Chemically stable Phenyl sorbent for ultra-low column bleed and long lifetimes
- 2.5 μm XP columns in ultra-low dispersion hardware
- Exceptional separation performance, robustness, and high throughput for HPLC
- Seamless transition to UPLC adoption
- Reversed phase separation mode
- pH range: 1 to 12
- Maximum pressure: 18000 psi (1240 Bar)
- Pore size: 130 Å
- Dimensions: 3 mm x 50 mm
- Units per package: 3 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.