Learn More
Waters Corp Waters Corporation Method Validation Kit, BEH Shield RP18, 2.5 μm, 130 Å, 3 mm X 150 mm
Supplier: Waters Corp 186006774
This XBridge BEH Shield RP18 Method Validation Kit includes three columns for HPLC method development. It offers alternative selectivity for phenolic compounds with its embedded carbamate group. The 2.5 μm XP columns provide exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC transition. Digital Certificates of Analysis are available online.
- Chemistry: C18
- Separation mode: Reversed phase
- Particle substrate: Hybrid
- pH range: 2 to 11
- Maximum pressure: 18000 psi (1240 Bar)
- Particle size: 2.5 μm
- Pore size: 130 Å
- System: UPLC, UHPLC
- Inner diameter: 3 mm
- Length: 150 mm
- Units per package: 3 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.