Learn More
Waters Corp XBridge Preparative Column, BEH Shield RP18, 10 x 100 mm, 5 μm
Supplier:  Waters Corp 186008169
Encompass_Preferred
The Waters XBridge Preparative Column with BEH Shield RP18 chemistry is designed for preparative chromatography, offering improved peak shape for basic compounds and direct scalability.
  • BEH Shield RP18 chemistry for enhanced performance
  • Improved peak shape for basic compounds
  • Direct scalability for preparative chromatography
  • 5 μm particle size
  • 10 x 100 mm dimensions
  • OBD Column type
Catalog No. 50-239-0639