Learn More
Waters Corp OBD Column, BEH Shield RP18, 10 x 50 mm, 5 μm
Supplier:  Waters Corp 186008168
Encompass_Preferred
The XBridge BEH Shield RP18 OBD Column is a preparative column featuring Ethylene-Bridged Hybrid (BEH Technology™). It offers direct scalability from analytical to preparative separations and provides excellent column lifetime and consistent results. Designed for a wide pH and temperature range, this column delivers good resolution and peak shape for diverse compounds, including basic analytes, maximizing recoveries for isolation processes.
  • Uniform, optimum packed bed density.
  • Direct scalability from analytical to preparative.
  • Excellent column lifetime and consistent performance.
  • Good resolution and peak shape, even for basic analytes.
  • Stable across a wide pH range and high temperatures.
  • Maximize recoveries of small molecules and biomolecules.
Catalog No. 50-239-2829