Learn More
Waters Corp Waters XBridge BEH Shield RP18 XP Column, 130Å, 2.5 μm, 2.1 mm × 30 mm
Supplier:  Waters Corp 176002562
Encompass_Preferred
XBridge BEH Shield RP18 columns provide alternate selectivity for HPLC method development, especially for phenolic compounds, due to an embedded carbamate group. These 2.5 μm XP columns deliver exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC transition. Features MaxPeak High-Performance Surface technology to prevent unwanted analyte-surface interactions and is compatible with 100% aqueous mobile phases.
  • Alternate selectivity for phenolic compounds.
  • Compatible with 100% aqueous mobile phases.
  • XP column hardware for separation, robustness, and throughput.
  • Seamless method transfer paths.
  • Protected by a VanGuard Cartridge for extended lifetime.
  • MaxPeak High-Performance Surface technology prevents analyte-surface interactions.
Catalog No. 50-179-221