Learn More
Waters Corp Waters Corporation Column, Shield RP18, 130Å, 2.5 μm, 4.6 mm x 50 mm
Supplier:  Waters Corp 186006061
Encompass_Preferred
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and offer a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds, compared to straight chain alkyl columns. These 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays, while providing a seamless transition path toward UPLC adoption.
  • Operates over a wide pH range with various solvents, offering alternate selectivity.
  • Facilitates superior base separation by decreasing silanol interactions.
  • Proprietary Shield Technology produces highly symmetrical, efficient peaks.
  • Offers greater pH stability, excellent efficiency, and symmetrical peak form.
Catalog No. 50-201-523