Learn More
Waters Corp HPLC Column, XBridge BEH Shield RP18, 2.5 μm, 2.1 mm x 75 mm, 130 Å
Supplier:  Waters Corp 186006054
Encompass_Preferred
Waters HPLC Column, XBridge BEH Shield RP18, 2.5 μm, 2.1 mm x 75 mm, 130 Å
Catalog No. 50-179-484