Learn More
Waters Corp Waters Column, Shield RP18, 2.5 μm, 3 mm × 100 mm
Supplier:  Waters Corp 186006059
Encompass_Preferred
XBridge BEH Shield RP18 columns offer alternate selectivity, particularly for phenolic compounds, through an embedded carbamate group. These 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance and throughput for HPLC assays, facilitating a seamless transition to UPLC. Shield Technology improves peak morphologies by decreasing interactions with basic analytes and producing highly symmetrical, efficient peaks.
  • Complements BEH C18 and C8 phases for HPLC method development.
  • Embedded carbamate group provides alternate selectivity for phenolic compounds.
  • XP ultra-low dispersion hardware ensures exceptional separation and throughput for HPLC.
  • Enables seamless transition to UPLC adoption.
  • Shield Technology improves peak morphologies and efficiency with basic analytes.
  • Certified quality parts ensure reliable instrument performance and precise results.
Catalog No. 50-179-489