Learn More
Waters Corp WATERS CORPORATION XBridge Column, C18, 130 Å, 2.5 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186006055
Encompass_Preferred
XBridge BEH Shield RP18 columns offer alternate selectivity for HPLC method development, especially for phenolic compounds. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns provide exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC transition.
  • Alternate selectivity for phenolic compounds via embedded carbamate group.
  • Exceptional separation, robustness, and throughput for HPLC assays.
  • Seamless transition path to UPLC adoption.
  • Superior peak shape for bases and compatibility with fully aqueous mobile phases.
  • Shield Technology ensures unique selectivity and stable, reproducible retention.
Catalog No. 50-179-485