Learn More
Waters Corp Waters XBridge BEH Shield RP18 Column, 130Å, 2.5 μm, 2.1 mm
Supplier:  Waters Corp 186006053
Encompass_Preferred
XBridge BEH Shield RP18 columns are designed for advanced HPLC method development, offering alternate selectivity for phenolic compounds due to an embedded carbamate group. These 2.5 μm eXtended Performance (XP) columns provide exceptional separation, robustness, and throughput, enabling a seamless transition to UPLC. Their Shield Technology reduces silanol interactions, ensuring symmetrical peak forms for basic analytes.
  • C18 chemistry for reversed phase separation
  • pH range of 2 to 11
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 2.5 μm; Pore size: 130 Å
Catalog No. 50-179-483