Learn More
Waters Corp Waters XBridge Column, BEH Shield RP18, 2.5 μm, 2.1 mm × 75 mm
Supplier:  Waters Corp 176002564
Encompass_Preferred
These XBridge BEH Shield RP18 columns are designed for HPLC method development, providing alternate selectivity for phenolic compounds with an embedded carbamate group. The 2.5 μm eXtended Performance (XP) columns, featuring ultra-low dispersion hardware, offer exceptional separation, robustness, and throughput for HPLC assays, and facilitate a smooth transition to UPLC. MaxPeak HPS technology helps prevent constant method reproduction and validation.
  • Offers alternate selectivity for phenolic compounds.
  • eXtended Performance (XP) format for exceptional separation, robustness, and throughput.
  • Features ultra-low dispersion hardware.
  • Designed for a seamless transition to UPLC.
  • MaxPeak HPS technology minimizes method reproduction and validation.
  • Compatible with a VanGuard Cartridge to prevent contaminants from fouling.
Catalog No. 50-179-223